Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4,7,10-Tetraazacyclododecane tetrahydrochloride, 98%

Chemical Identifiers

Chemical Name or Material 1,4,7,10-Tetraazacyclododecane tetrahydrochloride
CAS 10045-25-7
Molecular Formula C8H20N4·4HCl
Formula Weight 318.12
MDL Number MFCD00068484
UN Number UN3077
Synonym Cyclen tetrahydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2226801

  SDS
alfa aesar™
B2226801
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2226803

  SDS
alfa aesar™
B2226803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2226806

  SDS
alfa aesar™
B2226806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 318.12
Chemical Name or Material 1,4,7,10-Tetraazacyclododecane tetrahydrochloride
Assay 98%
Molecular Formula C8H20N4·4HCl
MDL Number MFCD00068484
CAS 10045-25-7
UN Number UN3077
Synonym Cyclen tetrahydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f-P273-P501a
Wear protective clothing.
Avoid release to the environment.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point >290°C
Odor Odorless
Quantity 0.25g
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P280f-P273-P501a
Wear protective clothing.
Avoid release to the environment.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.