missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic Acid, TCI America™

Manufacturer:  TCI America T18751G

 View more versions of this product

Catalog No. T18751G



Specifications

Specifications

1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic Acid
267°C
60239-18-1
MFCD00068657
WDLRUFUQRNWCPK-UHFFFAOYSA-N
2-[4,7,10-tris(carboxymethyl)-1,4,7,10-tetrazacyclododec-1-yl]acetic acid
121841
404.42
≥97.0% (T)
White-Yellow
1g
C16H28N4O8
DOTA
C1CN(CCN(CCN(CCN1CC(=O)O)CC(=O)O)CC(=O)O)CC(=O)O
404.42
CHEBI:61028
Crystal-Powder at 20°C

Chemical Identifiers

60239-18-1
404.42
WDLRUFUQRNWCPK-UHFFFAOYSA-N
121841
2-[4,7,10-tris(carboxymethyl)-1,4,7,10-tetrazacyclododec-1-yl]acetic acid
C16H28N4O8
MFCD00068657
DOTA
CHEBI:61028
C1CN(CCN(CCN(CCN1CC(=O)O)CC(=O)O)CC(=O)O)CC(=O)O
Safety and Handling

Safety and Handling

TSCA : No

Documents