Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4,6,7-Tetramethylnaphthalene, 98%

$349.42 - $349.42

Chemical Identifiers

Chemical Name or Material 1,4,6,7-Tetramethylnaphthalene
CAS 13764-18-6
Molecular Formula C14H16
Formula Weight 184.28
MDL Number MFCD00049173
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2135106

 
alfa aesar™
B2135106
5g Each for $349.42
Description & Specifications

Specifications

Formula Weight 184.28
Chemical Name or Material 1,4,6,7-Tetramethylnaphthalene
Assay 98%
Molecular Formula C14H16
MDL Number MFCD00049173
CAS 13764-18-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 63° to 64°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.