missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4,5,8-Naphthalenetetracarboxylic Acid (contains Monoanhydride), TCI America™

$149.72 - $791.41

Chemical Identifiers

CAS 128-97-2
Molecular Formula C14H8O8
Molecular Weight (g/mol) 304.21
InChI Key OLAPPGSPBNVTRF-UHFFFAOYSA-N
PubChem CID 31422
IUPAC Name naphthalene-1,4,5,8-tetracarboxylic acid
SMILES C1=CC(=C2C(=CC=C(C2=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0770250G View Documents TCI America
N0770250G
250g Each for $791.41
 
N077025G View Documents TCI America
N077025G
25g Each for $149.72
 
Specifications

Chemical Identifiers

128-97-2
304.21
31422
C1=CC(=C2C(=CC=C(C2=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O
C14H8O8
OLAPPGSPBNVTRF-UHFFFAOYSA-N
naphthalene-1,4,5,8-tetracarboxylic acid

Specifications

Yellow
128-97-2
OLAPPGSPBNVTRF-UHFFFAOYSA-N
naphthalene-1,4,5,8-tetracarboxylic acid
31422
Crystal-Powder at 20°C
1,4,5,8-Naphthalenetetracarboxylic Acid (contains Monoanhydride)
250g
C14H8O8
C1=CC(=C2C(=CC=C(C2=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O
304.21
304.21
≥60.0% (NMR)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1009

RTECSNumber : QK3690000

TSCA : Yes

Recommended Storage : Refrigerator