missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4,5,8-Naphthalenetetracarboxdiimide, TCI America™

$275.00 - $944.19

Chemical Identifiers

CAS 5690-24-4
Molecular Formula C14H6N2O4
Molecular Weight (g/mol) 266.212
MDL Number MFCD00047649
InChI Key BODUWJSFPLUDMP-UHFFFAOYSA-N
PubChem CID 79771
SMILES C1=CC2=C3C(=CC=C4C3=C1C(=O)NC4=O)C(=O)NC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N05361G View Documents TCI America
N05361G
1g Each for $275.00
 
N05365G View Documents TCI America
N05365G
5g Each for $944.19
 
Specifications

Chemical Identifiers

5690-24-4
266.212
BODUWJSFPLUDMP-UHFFFAOYSA-N
C1=CC2=C3C(=CC=C4C3=C1C(=O)NC4=O)C(=O)NC2=O
C14H6N2O4
MFCD00047649
79771

Specifications

Undesignated
5690-24-4
MFCD00047649
C1=CC2=C3C(=CC=C4C3=C1C(=O)NC4=O)C(=O)NC2=O
79771
Crystal-Powder at 20°C
1,4,5,8-Naphthalenetetracarboxdiimide
1g
C14H6N2O4
BODUWJSFPLUDMP-UHFFFAOYSA-N
266.212
266.21
≥97.0% (N)
Safety and Handling

Safety and Handling

TSCA : No