Please login to your online account to display your discounted pricing

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine, ≥97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC30139DA

 View more versions of this product

Catalog No. CC30139DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 273.18
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
Molecular Formula C16H24BNO2
CAS 852227-90-8
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD08060504
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed. Call a POISON CENTER or doctor/physician if you feel unwell.

Specifications

Chemical Name or Material 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
CAS 852227-90-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H24BNO2
Formula Weight 273.18
MDL Number MFCD08060504
Accessories

GHS Signal Word: Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

SDS & Safety Documents