missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine, ≥97%, Maybridge

Chemical Identifiers

CAS 852227-90-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C16H24BNO2
Formula Weight 273.18
MDL Number MFCD08060504
Chemical Name or Material 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC30139CB SDS Fisher Scientific Company
CC30139CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30139DA SDS Fisher Scientific Company
CC30139DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

852227-90-8
100.0
C16H24BNO2
MFCD08060504
97%
96.5
97%
273.18
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine

Specifications

273.18
100.0
97%
96.5
852227-90-8
97%
250mg
C16H24BNO2
MFCD08060504
Amber Glass Bottle
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine

Safety and Handling

GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning