Please login to your online account to display your discounted pricing

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC33939DA

 View more versions of this product

Catalog No. CC33939DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 287.21
Quantity 1g
Assay 97%
Chemical Name or Material 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
Molecular Formula C17H26BNO2
CAS 852227-96-4
Packaging Amber Glass Bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
Assay 97%
Assay Percent Range 97%
Molecular Formula C17H26BNO2
Formula Weight 287.21