missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-[3-(Trimethoxysilyl)propyl]urea 94.0+%, TCI America™
Supplier: TCI America T191525G
Specifications
1-[3-(Trimethoxysilyl)propyl]urea | |
Yellow | |
MFCD00216654 | |
(3-Ureidopropyl)trimethoxysilane | |
CO[Si](CCCNC(N)=O)(OC)OC | |
222.32 | |
222.32 | |
Liquid |
23843-64-3 | |
C7H18N2O4Si | |
25 g | |
LVACOMKKELLCHJ-UHFFFAOYSA-N | |
[3-(trimethoxysilyl)propyl]urea | |
90280 | |
≥94.0% (N) |
Chemical Identifiers
23843-64-3 | |
222.32 | |
LVACOMKKELLCHJ-UHFFFAOYSA-N | |
90280 | |
CO[Si](CCCNC(N)=O)(OC)OC |
C7H18N2O4Si | |
MFCD00216654 | |
(3-Ureidopropyl)trimethoxysilane | |
[3-(trimethoxysilyl)propyl]urea |
Safety and Handling
TSCA : Yes