missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[3-(Trimethoxysilyl)propyl]urea, TCI America™

Manufacturer:  TCI America T1915250G

 View more versions of this product

Catalog No. T1915250G



Specifications

Specifications

1-[3-(Trimethoxysilyl)propyl]urea
250g
C7H18N2O4Si
(3-Ureidopropyl)trimethoxysilane
CO[Si](CCCNC(=O)N)(OC)OC
222.316
222.32
≥94.0% (N)
Yellow
23843-64-3
MFCD00216654
LVACOMKKELLCHJ-UHFFFAOYSA-N
3-trimethoxysilylpropylurea
90280
Clear Liquid at 20°C

Chemical Identifiers

23843-64-3
222.316
LVACOMKKELLCHJ-UHFFFAOYSA-N
90280
CO[Si](CCCNC(=O)N)(OC)OC
C7H18N2O4Si
MFCD00216654
(3-Ureidopropyl)trimethoxysilane
3-trimethoxysilylpropylurea
Safety and Handling

Safety and Handling

TSCA : Yes

Documents