missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[3-(Trifluoromethyl)phenylsulfonylamino]cyclohexanecarboxylic acid, 96%, Alfa Aesar™

$39.95 - $109.03

Chemical Identifiers

CAS 690645-95-5
Molecular Formula C14H16F3NO4S
Molecular Weight (g/mol) 351.34
MDL Number MFCD05866546
InChI Key DDEMDTVJTHGHDA-UHFFFAOYSA-N
Synonym 1-3-trifluoromethyl benzenesulfonamido cyclohexane-1-carboxylic acid
PubChem CID 2241577
IUPAC Name 1-[[3-(trifluoromethyl)phenyl]sulfonylamino]cyclohexane-1-carboxylic acid
SMILES C1CCC(CC1)(C(=O)O)NS(=O)(=O)C2=CC=CC(=C2)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH34351MD View Documents Alfa Aesar
H34351MD
250mg Each for $39.95
 
AAH3435103 View Documents Alfa Aesar
H3435103
1g Each for $109.03
 
Specifications

Chemical Identifiers

690645-95-5
351.34
DDEMDTVJTHGHDA-UHFFFAOYSA-N
2241577
C1CCC(CC1)(C(=O)O)NS(=O)(=O)C2=CC=CC(=C2)C(F)(F)F
C14H16F3NO4S
MFCD05866546
1-3-trifluoromethyl benzenesulfonamido cyclohexane-1-carboxylic acid
1-[[3-(trifluoromethyl)phenyl]sulfonylamino]cyclohexane-1-carboxylic acid

Specifications

164°C to 165°C
690645-95-5
MFCD05866546
DDEMDTVJTHGHDA-UHFFFAOYSA-N
1-[[3-(trifluoromethyl)phenyl]sulfonylamino]cyclohexane-1-carboxylic acid
2241577
96%
250mg
C14H16F3NO4S
1-3-trifluoromethyl benzenesulfonamido cyclohexane-1-carboxylic acid
C1CCC(CC1)(C(=O)O)NS(=O)(=O)C2=CC=CC(=C2)C(F)(F)F
351.34
351.35
1-[3-(Trifluoromethyl)phenylsulfonylamino]cyclohexanecarboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning