Please login to your online account to display your discounted pricing

1,3-Thiazole-2-carboxylic acid, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC09501DA

 View more versions of this product

Catalog No. CC09501DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 129.14
Quantity 1g
Assay 95%
CAS Max % 100.0
MDL Number MFCD02094169
CAS Min % 94.5
Chemical Name or Material 1, 3-Thiazole-2-carboxylic acid
Packaging Amber Glass Bottle
Molecular Formula C4H3NO2S
CAS 14190-59-1
Assay Percent Range 95%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

Specifications

Chemical Name or Material 1, 3-Thiazole-2-carboxylic acid
CAS 14190-59-1
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C4H3NO2S
Formula Weight 129.14
MDL Number MFCD02094169
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

SDS & Safety Documents