missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Thiazol-2-ylmethanol, 97+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC09509EA
Specifications
1, 3-Thiazol-2-ylmethanol | |
97% | |
C4H5NOS | |
10g | |
JNHDLNXNYPLBMJ-UHFFFAOYSA-N | |
1,3-thiazol-2-ylmethanol | |
2795213 | |
97% |
14542-12-2 | |
Amber Glass Bottle | |
MFCD06200855 | |
thiazol-2-ylmethanol, 2-thiazolemethanol, thiazol-2-yl-methanol, 2-hydroxymethylthiazole, 2-hydroxymethyl thiazole, 2-hydroxymehtylthiazole, 1,3-thiazol-2-yl methanol, thiazol-2-yl methanol, thiazole-2-methanol, 2-thiazolylmethanol | |
OCC1=NC=CS1 | |
115.15 | |
115.16 |
Chemical Identifiers
14542-12-2 | |
115.15 | |
JNHDLNXNYPLBMJ-UHFFFAOYSA-N | |
2795213 | |
OCC1=NC=CS1 |
C4H5NOS | |
MFCD06200855 | |
thiazol-2-ylmethanol, 2-thiazolemethanol, thiazol-2-yl-methanol, 2-hydroxymethylthiazole, 2-hydroxymethyl thiazole, 2-hydroxymehtylthiazole, 1,3-thiazol-2-yl methanol, thiazol-2-yl methanol, thiazole-2-methanol, 2-thiazolylmethanol | |
1,3-thiazol-2-ylmethanol |
Safety and Handling
Warning