Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3-tert-Butoxyphenyl)homopiperazine monohydrochloride, 98%

$430.24 - $430.24

Chemical Identifiers

Chemical Name or Material 1-(3-tert-Butoxyphenyl)homopiperazine monohydrochloride
CAS 934992-04-8
Molecular Formula C15H24N2O·HCl
Formula Weight 284.83g/mol
MDL Number MFCD16172200
Synonym 3-BOPH·HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51752ME

 
alfa aesar™
H51752-ME
500mg Each for $430.24
Description & Specifications

Specifications

Quantity 500mg
Melting Point 143° to 147°C
Quantity 500mg
Chemical Name or Material 1-(3-tert-Butoxyphenyl)homopiperazine monohydrochloride
CAS 934992-04-8
Assay 98%
Molecular Formula C15H24N2O·HCl
Formula Weight 284.83g/mol
MDL Number MFCD16172200
Synonym 3-BOPH·HCl