missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(3-Sulfopropyl)pyridinium Hydroxide Inner Salt 98.0+%, TCI America™

Manufacturer:  TCI America S039125G

 View more versions of this product

Catalog No. S039125G



Specifications

Specifications

1-(3-Sulfopropyl)pyridinium Hydroxide Inner Salt
279°C
3 to 6 (10% H2O soln. 20°C)
C8H11NO3S
3-pyridin-1-ium-1-yl propane-1-sulfonate, ndsb-201, 1-3-sulphonatopropyl pyridinium, unii-qn4i6ai9ek, 3-1-pyridinio-1-propanesulfonate, 1-pyridiniumpropane-3-sulfonate, qn4i6ai9ek, 1-3-sulfopropyl pyridinium hydroxide, inner salt, pyridinium, 1-3-sulfopropyl-, inner salt, 1-3-sulfonatopropyl pyridin-1-ium
C1=CC=[N+](C=C1)CCCS(=O)(=O)[O-]
201.24
201.24
≥98.0% (HPLC,N)
White
25g
15471-17-7
MFCD00064468
REEBJQTUIJTGAL-UHFFFAOYSA-N
3-pyridin-1-ium-1-ylpropane-1-sulfonate
84929
Crystal-Powder at 20°C

Chemical Identifiers

15471-17-7
201.24
REEBJQTUIJTGAL-UHFFFAOYSA-N
84929
C1=CC=[N+](C=C1)CCCS(=O)(=O)[O-]
C8H11NO3S
MFCD00064468
3-pyridin-1-ium-1-yl propane-1-sulfonate, ndsb-201, 1-3-sulphonatopropyl pyridinium, unii-qn4i6ai9ek, 3-1-pyridinio-1-propanesulfonate, 1-pyridiniumpropane-3-sulfonate, qn4i6ai9ek, 1-3-sulfopropyl pyridinium hydroxide, inner salt, pyridinium, 1-3-sulfopropyl-, inner salt, 1-3-sulfonatopropyl pyridin-1-ium
3-pyridin-1-ium-1-ylpropane-1-sulfonate
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0758&(5)-3704

TSCA : Yes

Documents