missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Propanesultone, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1192318

 View more versions of this product

Catalog No. AAA1192318


 

Description

Description

Applications
1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.

Solubility
Soluble in water (100 mg/ml), chloroform, esters, methanol, and aromatic hydrocartions.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
Specifications

Specifications

1, 3-Propanesultone
1.392
C3H6O3S
C1COS(=O)(=O)C1
122.138
CHEBI:82370
50g
1120-71-4
99%
FSSPGSAQUIYDCN-UHFFFAOYSA-N
oxathiolane 2,2-dioxide
14264
122.14

Chemical Identifiers

1120-71-4
122.138
14264
C3H6O3S
FSSPGSAQUIYDCN-UHFFFAOYSA-N
CHEBI:82370
Safety and Handling

Safety and Handling

storageNote1 : Moisture Sensitive

SDS Documents