Learn More
1,3-Propanedithiol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131410500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 3-Propanedithiol | |
97.5 | |
-79°C | |
1.0700g/mL | |
58°C | |
97.5% min. (GC) | |
C3H8S2 | |
HS(CH2)3SH | |
50 mL | |
01,956; 04,413; 06,493; 09,394; 13,261 | |
15, 7915 | |
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,ether,chloroform and,benzene | |
C(CS)CS | |
108.22 | |
CHEBI:44864 | |
98% |
109-80-8 | |
100.0 | |
Yellow | |
169°C | |
Authentic | |
Glass bottle | |
1.5380 to 1.542 | |
MFCD00004904 | |
01, 476 | |
1.07 | |
1,3-propanedithiol, 1,3-dimercaptopropane, trimethylene dimercaptan, trimethylenedithiol, dithiotrimethyleneglycol, trimethylenedithioglycol, 1,3-propanedimercaptan, unii-r4luj82u52, fema no. 3588, 1,3-propane dithiol | |
ZJLMKPKYJBQJNH-UHFFFAOYSA-N | |
propane-1,3-dithiol | |
8013 | |
108.22 | |
Liquid |
Chemical Identifiers
109-80-8 | |
108.22 | |
ZJLMKPKYJBQJNH-UHFFFAOYSA-N | |
8013 | |
propane-1,3-dithiol |
C3H8S2 | |
MFCD00004904 | |
1,3-propanedithiol, 1,3-dimercaptopropane, trimethylene dimercaptan, trimethylenedithiol, dithiotrimethyleneglycol, trimethylenedithioglycol, 1,3-propanedimercaptan, unii-r4luj82u52, fema no. 3588, 1,3-propane dithiol | |
CHEBI:44864 | |
C(CS)CS |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
GHS Signal Word: Danger
EINECSNumber : 203-706-9
RUO – Research Use Only