Please login to your online account to display your discounted pricing

1,3-Propanediol di-p-tosylate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Propanediol di-p-tosylate
CAS 5469-66-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C17H20O6S2
Linear Formula CH3C6H4SO3(CH2)3O3SC6H4CH3
Formula Weight 384.47
MDL Number MFCD00013392
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC373090250

  SDS
thermo fisher scientific
AC373090250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥98.5%
Formula Weight 384.47
Quantity 25g
Color White to off-white
Melting Point 90° to 93°C
CAS Max % 100.0
Molecular Formula C17H20O6S2
Linear Formula CH3C6H4SO3(CH2)3O3SC6H4CH3
MDL Number MFCD00013392
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Assay Percent Range 99%
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 1, 3-Propanediol di-p-tosylate
CAS 5469-66-9
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Alkylation reagent

Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.