missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Propanediamine-N,N,N',N'-tetraacetic Acid 98.0+%, TCI America™
$43.13 - $272.08
Chemical Identifiers
CAS | 1939-36-2 |
---|---|
Molecular Formula | C11H18N2O8 |
Molecular Weight (g/mol) | 306.27 |
MDL Number | MFCD00210039 |
InChI Key | DMQQXDPCRUGSQB-UHFFFAOYSA-N |
PubChem CID | 80296 |
IUPAC Name | 2-({3-[bis(carboxymethyl)amino]propyl}(carboxymethyl)amino)acetic acid |
SMILES | OC(=O)CN(CCCN(CC(O)=O)CC(O)=O)CC(O)=O |
Chemical Identifiers
1939-36-2 | |
306.27 | |
DMQQXDPCRUGSQB-UHFFFAOYSA-N | |
2-({3-[bis(carboxymethyl)amino]propyl}(carboxymethyl)amino)acetic acid |
C11H18N2O8 | |
MFCD00210039 | |
80296 | |
OC(=O)CN(CCCN(CC(O)=O)CC(O)=O)CC(O)=O |
Specifications
1939-36-2 | |
White | |
MFCD00210039 | |
Trimethylenediamine-N,N,N′C,N′C-tetraacetic Acid | |
OC(=O)CN(CCCN(CC(O)=O)CC(O)=O)CC(O)=O | |
306.27 | |
306.27 | |
Crystalline Powder |
227°C | |
C11H18N2O8 | |
25 g | |
DMQQXDPCRUGSQB-UHFFFAOYSA-N | |
2-({3-[bis(carboxymethyl)amino]propyl}(carboxymethyl)amino)acetic acid | |
80296 | |
≥98.0% (T) | |
1,3-Propanediamine-N,N,N′,N′-tetraacetic Acid |
Safety and Handling
EINECSNumber : (2)-3632
RTECSNumber : MC1082500
TSCA : Yes