missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Propanediamine-N,N,N′,N′-tetraacetic Acid, TCI America™

Manufacturer:  TCI America P160525G

 View more versions of this product

Catalog No. P160525G



Specifications

Specifications

1,3-Propanediamine-N,N,N′,N′-tetraacetic Acid
≥98.0% (T)
Trimethylenediamine-N,N,N′C,N′C-tetraacetic Acid
C(CN(CC(=O)O)CC(=O)O)CN(CC(=O)O)CC(=O)O
306.271
306.27
White
25g
1939-36-2
C11H18N2O8
DMQQXDPCRUGSQB-UHFFFAOYSA-N
2-[3-[bis(carboxymethyl)amino]propyl-(carboxymethyl)amino]acetic acid
80296
Crystal-Powder at 20°C
227°C

Chemical Identifiers

1939-36-2
306.271
Trimethylenediamine-N,N,N′C,N′C-tetraacetic Acid
C11H18N2O8
DMQQXDPCRUGSQB-UHFFFAOYSA-N
80296
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3632

RTECSNumber : MC1082500

TSCA : Yes

Documents