missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Phenylenediacetic acid, 97%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 130500050

 View more versions of this product

Catalog No. AC130500050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
194.19
White to light yellow
173° to 177°C
100.0
Glass bottle
97%
C10H10O4
Authentic
Fine Crystalline Powder
5g
09, 874
96.0
19806-17-8
C6H4(CH2CO2H)2
1, 3-Phenylenediacetic acid
MFCD00004341

Chemical Identifiers

19806-17-8
100.0
C10H10O4
194.19
1, 3-Phenylenediacetic acid
96.0
97%
C6H4(CH2CO2H)2
MFCD00004341
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.