Please login to your online account to display your discounted pricing

1,3-Phenylenediacetic acid, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC130500250

 View more versions of this product

Catalog No. AC130500250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Formula Weight 194.19
Assay ≥96.0%
Quantity 25g
Color White to light yellow
Melting Point 173° to 177°C
Beilstein 09, 874
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
CAS 19806-17-8
Linear Formula C6H4(CH2CO2H)2
Chemical Name or Material 1, 3-Phenylenediacetic acid
Molecular Formula C10H10O4
Assay Percent Range 97%
MDL Number MFCD00004341
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1, 3-Phenylenediacetic acid
CAS 19806-17-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H10O4
Linear Formula C6H4(CH2CO2H)2
Formula Weight 194.19
MDL Number MFCD00004341
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents