missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Pentadiene (cis- and trans- mixture) (stabilized with TBC), TCI America™

Manufacturer:  TCI America P184125ML

 View more versions of this product

Catalog No. P184125ML



Specifications

Specifications

1,3-Pentadiene (cis- and trans- mixture) (stabilized with TBC)
Yellow
504-60-9
MFCD00009295
1,3-pentadiene, trans-1,3-pentadiene, piperylene, trans-piperylene, e-1,3-pentadiene, 1-methylbutadiene, 3e-penta-1,3-diene, trans-1-methylbutadiene, 1,3-pentadiene, 3e, trans-penta-1,3-diene
CC=CC=C
68.119
CHEBI:74165
Clear Liquid at 20°C
42°C
25mL
C5H8
3295
PMJHHCWVYXUKFD-SNAWJCMRSA-N
(3E)-penta-1,3-diene
62204
68.12
≥96.0% (GC)

Chemical Identifiers

504-60-9
68.119
PMJHHCWVYXUKFD-SNAWJCMRSA-N
62204
(3E)-penta-1,3-diene
C5H8
MFCD00009295
1,3-pentadiene, trans-1,3-pentadiene, piperylene, trans-piperylene, e-1,3-pentadiene, 1-methylbutadiene, 3e-penta-1,3-diene, trans-1-methylbutadiene, 1,3-pentadiene, 3e, trans-penta-1,3-diene
CHEBI:74165
CC=CC=C
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0020

RTECSNumber : RZ2464000

TSCA : Yes

Recommended Storage : Refrigerator

Documents