Please login to your online account to display your discounted pricing

1-(3-Methylpyrazin-2-yl)ethan-1-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-(3-Methylpyrazin-2-yl)ethan-1-one
CAS 23787-80-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H8N2O
Formula Weight 136.15
MDL Number MFCD00014612
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01015EA

 
fisher scientific company
SB01015EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 136.15
Quantity 10g
Assay 97%
Density 1.1100g/mL
CAS Min % 97.0
Molecular Formula C7H8N2O
Chemical Name or Material 1-(3-Methylpyrazin-2-yl)ethan-1-one
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD00014612
CAS 23787-80-6