Please login to your online account to display your discounted pricing

1-(3-Methylpyrazin-2-yl)ethan-1-one, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SB01015EA

 View more versions of this product

Catalog No. SB01015EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 136.15
Quantity 10g
Assay 97%
Density 1.1100g/mL
CAS Min % 97.0
Molecular Formula C7H8N2O
Chemical Name or Material 1-(3-Methylpyrazin-2-yl)ethan-1-one
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD00014612
CAS 23787-80-6

Specifications

Chemical Name or Material 1-(3-Methylpyrazin-2-yl)ethan-1-one
CAS 23787-80-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H8N2O
Formula Weight 136.15
MDL Number MFCD00014612