missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(3-Methylphenyl)piperazine, 99%, ACROS Organics™
$58.32 - $139.11
Chemical Identifiers
CAS | 41186-03-2 |
---|---|
Molecular Formula | C11H16N2 |
Molecular Weight (g/mol) | 176.26 |
MDL Number | MFCD00040734 |
InChI Key | JIWHIRLNKIUYSM-UHFFFAOYSA-N |
Synonym | 1-3-methylphenyl piperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl piperazine, 1-m-tolyl-piperazine, 1-m-tolyl-piperzine, 1-m-tolylpiperazine, 3-methylphenyl piperazine, 4-m-tolyl-piperazine, 4-m-tolylpiperazine, piperazine, 1-3-methylphenyl |
PubChem CID | 83111 |
IUPAC Name | 1-(3-methylphenyl)piperazine |
SMILES | CC1=CC(=CC=C1)N2CCNCC2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC345950010
|
Thermo Fisher Scientific
345950010 |
1g | Glass bottle |
Each for $58.32
|
|
AC345950050
|
Thermo Fisher Scientific
345950050 |
5g | Glass bottle |
Each for $139.11
|
|
Chemical Identifiers
41186-03-2 | |
176.26 | |
JIWHIRLNKIUYSM-UHFFFAOYSA-N | |
83111 | |
CC1=CC(=CC=C1)N2CCNCC2 |
C11H16N2 | |
MFCD00040734 | |
1-3-methylphenyl piperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl piperazine, 1-m-tolyl-piperazine, 1-m-tolyl-piperzine, 1-m-tolylpiperazine, 3-methylphenyl piperazine, 4-m-tolyl-piperazine, 4-m-tolylpiperazine, piperazine, 1-3-methylphenyl | |
1-(3-methylphenyl)piperazine |
Specifications
Authentic | |
1.5755 to 1.5785 | |
1g | |
98.5 | |
98.5% min. (GC) | |
MFCD00040734 | |
JIWHIRLNKIUYSM-UHFFFAOYSA-N | |
1-(3-methylphenyl)piperazine | |
83111 | |
Viscous Liquid | |
1-(3-Methylphenyl)piperazine, 99% |
Glass bottle | |
Colorless to Yellow | |
41186-03-2 | |
100.0 | |
C11H16N2 | |
1-3-methylphenyl piperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl piperazine, 1-m-tolyl-piperazine, 1-m-tolyl-piperzine, 1-m-tolylpiperazine, 3-methylphenyl piperazine, 4-m-tolyl-piperazine, 4-m-tolylpiperazine, piperazine, 1-3-methylphenyl | |
CC1=CC(=CC=C1)N2CCNCC2 | |
176.26 | |
176.26 | |
99% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
EINECSNumber : 255-251-0
TSCA : TSCA