Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3-Methylphenyl)homopiperazine monohydrochloride, 98%

$430.24 - $430.24

Chemical Identifiers

Chemical Name or Material 1-(3-Methylphenyl)homopiperazine monohydrochloride
CAS 934991-97-6
Molecular Formula C12H18N2·HCl
Formula Weight 226.75g/mol
MDL Number MFCD08436120
Synonym 3-MPHP·HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51700ME

 
alfa aesar™
H51700-ME
500mg Each for $430.24
Description & Specifications

Specifications

Quantity 500mg
Melting Point 136° to 140°C
Quantity 500mg
Chemical Name or Material 1-(3-Methylphenyl)homopiperazine monohydrochloride
CAS 934991-97-6
Assay 98%
Molecular Formula C12H18N2·HCl
Formula Weight 226.75g/mol
MDL Number MFCD08436120
Synonym 3-MPHP·HCl