Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3-Methoxyphenyl)piperazine dihydrochloride, 97%

$105.78 - $388.74

Chemical Identifiers

Chemical Name or Material 1-(3-Methoxyphenyl)piperazine dihydrochloride
CAS 6968-76-9
Molecular Formula C11H16N2O·2HCl
Formula Weight 265.18
MDL Number MFCD00035285
Beilstein 5647713
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0216406

  SDS
alfa aesar™
L0216406
5g Each for $105.78

AAL0216414

  SDS
alfa aesar™
L0216414
25g Each for $388.74
Description & Specifications

Specifications

Formula Weight 265.18
Chemical Name or Material 1-(3-Methoxyphenyl)piperazine dihydrochloride
Assay 97%
Health Hazard 1 Warning
MDL Number MFCD00035285
Molecular Formula C11H16N2O·2HCl
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈214°C
Quantity 5g
Beilstein 5647713
CAS 6968-76-9
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
1-(3-Methoxyphenyl)piperazine dihydrochloride is used as an arylpiperazine compound which is an important intermediate in the preparation of various pharmaceuticals, used to treat depression and post-traumatic stress.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.