Learn More
1-(3-Methoxyphenyl)piperazine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5545014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-(3-Methoxyphenyl)piperazine | |
1.114 | |
91°C (196°F) | |
1.5815 | |
25 g | |
1-3-methoxyphenyl piperazine, n-3-methoxyphenyl piperazine, 1-3-methoxyphenyl-piperazine, unii-6lcp52962g, 1-3-methoxy-phenyl-piperazine, chembl59597, piperazine, 1-3-methoxyphenyl, 3-methoxy-1-piperazinylbenzene, pubchem8585, 3-meopp | |
COC1=CC=CC(=C1)N2CCNCC2 | |
192.262 | |
192.26 |
16015-71-7 | |
150°C (0.5 mmHg) | |
C11H16N2O | |
MFCD00040733 | |
UN3267 | |
PZIBVWUXWNYTNL-UHFFFAOYSA-N | |
1-(3-methoxyphenyl)piperazine | |
81430 | |
95% |
Chemical Identifiers
16015-71-7 | |
192.262 | |
PZIBVWUXWNYTNL-UHFFFAOYSA-N | |
81430 | |
COC1=CC=CC(=C1)N2CCNCC2 |
C11H16N2O | |
MFCD00040733 | |
1-3-methoxyphenyl piperazine, n-3-methoxyphenyl piperazine, 1-3-methoxyphenyl-piperazine, unii-6lcp52962g, 1-3-methoxy-phenyl-piperazine, chembl59597, piperazine, 1-3-methoxyphenyl, 3-methoxy-1-piperazinylbenzene, pubchem8585, 3-meopp | |
1-(3-methoxyphenyl)piperazine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 240-154-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only