missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(3-Methoxyphenoxy)-2-propanol, TCI America™

Manufacturer:  TCI America M106125G

 View more versions of this product

Catalog No. M106125G



Specifications

Specifications

1-(3-Methoxyphenoxy)-2-propanol
Yellow
382141-68-6
MFCD00191540
GUOGSANXPRRCMX-UHFFFAOYSA-N
1-(3-methoxyphenoxy)propan-2-ol
22036449
Clear Liquid at 20°C
110°C
25g
C10H14O3
3-(2-Hydroxypropoxy)anisole
CC(COC1=CC=CC(=C1)OC)O
182.219
182.22
≥90.0% (GC)

Chemical Identifiers

382141-68-6
182.219
GUOGSANXPRRCMX-UHFFFAOYSA-N
22036449
CC(COC1=CC=CC(=C1)OC)O
C10H14O3
MFCD00191540
3-(2-Hydroxypropoxy)anisole
1-(3-methoxyphenoxy)propan-2-ol
Safety and Handling

Safety and Handling

TSCA : No

Documents