missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(3-Methoxyphenoxy)-2-propanol, TCI America™

$189.45 - $310.14

Chemical Identifiers

CAS 382141-68-6
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00191540
InChI Key GUOGSANXPRRCMX-UHFFFAOYSA-N
Synonym 3-(2-Hydroxypropoxy)anisole
PubChem CID 22036449
IUPAC Name 1-(3-methoxyphenoxy)propan-2-ol
SMILES CC(COC1=CC=CC(=C1)OC)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M10615G View Documents TCI America
M10615G
5g Each for $189.45
 
M106125G View Documents TCI America
M106125G
25g Each for $310.14
 
Specifications

Chemical Identifiers

382141-68-6
182.219
GUOGSANXPRRCMX-UHFFFAOYSA-N
22036449
CC(COC1=CC=CC(=C1)OC)O
C10H14O3
MFCD00191540
3-(2-Hydroxypropoxy)anisole
1-(3-methoxyphenoxy)propan-2-ol

Specifications

110°C
5g
C10H14O3
3-(2-Hydroxypropoxy)anisole
CC(COC1=CC=CC(=C1)OC)O
182.219
182.22
≥90.0% (GC)
Yellow
382141-68-6
MFCD00191540
GUOGSANXPRRCMX-UHFFFAOYSA-N
1-(3-methoxyphenoxy)propan-2-ol
22036449
Clear Liquid at 20°C
1-(3-Methoxyphenoxy)-2-propanol
Safety and Handling

Safety and Handling

TSCA : No