missing translation for 'onlineSavingsMsg'
Learn More

1,3-Indanedione, 97%

CAS: 606-23-5 | C9H6O2 | 146.15 g/mol

$248.52 - $248.52

Chemical Identifiers

CAS 606-23-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
InChI Key UHKAJLSKXBADFT-UHFFFAOYSA-N
Synonym 1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g
PubChem CID 11815
ChEBI CHEBI:78877
IUPAC Name indene-1,3-dione
SMILES C1C(=O)C2=CC=CC=C2C1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122090250
SDS View Documents
Thermo Scientific Chemicals
122090250
25 g Glass bottle
Each for $248.52
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

606-23-5
146.15
1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g
CHEBI:78877
C1C(=O)C2=CC=CC=C2C1=O
C9H6O2
UHKAJLSKXBADFT-UHFFFAOYSA-N
11815
indene-1,3-dione

Specifications

606-23-5
Beige to White
97%
C9H6O2
07,694
UHKAJLSKXBADFT-UHFFFAOYSA-N
indene-1,3-dione
11815
146.15
Powder
129.0°C to 132.0°C
Authentic
Glass bottle
25 g
1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g
C1C(=O)C2=CC=CC=C2C1=O
146.15
CHEBI:78877
97%
1, 3-Indanedione
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 210-109-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only