Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3-Fluorophenyl)homopiperazine monohydrochloride, 98%

$668.05 - $668.05

Chemical Identifiers

Chemical Name or Material 1-(3-Fluorophenyl)homopiperazine monohydrochloride
CAS 934991-99-8
Molecular Formula C11H15FN2·HCl
Formula Weight 230.71
MDL Number MFCD09953903
Synonym 3-FPHP·HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51686ME

 
alfa aesar™
H51686ME
500mg Each for $668.05
Description & Specifications

Specifications

Formula Weight 230.71
Chemical Name or Material 1-(3-Fluorophenyl)homopiperazine monohydrochloride
Assay 98%
Molecular Formula C11H15FN2·HCl
MDL Number MFCD09953903
CAS 934991-99-8
Synonym 3-FPHP·HCl
Melting Point 187° to 191°C
Quantity 500mg