Please login to your online account to display your discounted pricing

1-(3-Fluorophenyl)-2-thiourea, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 1-(3-Fluorophenyl)-2-thiourea
Assay Percent Range 98%
Molecular Formula C7H7FN2S
Formula Weight 170.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372510050

  SDS
thermo fisher scientific
AC372510050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC372510010

  SDS
thermo fisher scientific
AC372510010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.21
Quantity 5g
Chemical Name or Material 1-(3-Fluorophenyl)-2-thiourea
Assay Percent Range 98%
Molecular Formula C7H7FN2S
CAS 458-05-9
Packaging Glass bottle