missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3-Fluorobenzyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5529503

 View more versions of this product

Catalog No. AAH5529503



Specifications

Specifications

1-(3-Fluorobenzyl)piperazine
1.5290
Colorless to Orange
55513-19-4
MFCD02242870
ITHBJSRWFNLKIH-UHFFFAOYSA-N
1-[(3-fluorophenyl)methyl]piperazine
903813
Liquid
1.106g/mL
281°C
1g
C11H15FN2
1-3-fluoro-benzyl-piperazine
C1CN(CCN1)CC2=CC(=CC=C2)F
194.253
194.25
97%

Chemical Identifiers

55513-19-4
194.253
ITHBJSRWFNLKIH-UHFFFAOYSA-N
903813
C1CN(CCN1)CC2=CC(=CC=C2)F
C11H15FN2
MFCD02242870
1-3-fluoro-benzyl-piperazine
1-[(3-fluorophenyl)methyl]piperazine
Documents