missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3-Fluorobenzyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5529514

 View more versions of this product

Catalog No. AAH5529514



Specifications

Specifications

1-(3-Fluorobenzyl)piperazine
>110°C (230°F)
281°C
25g
C11H15FN2
1-3-fluoro-benzyl-piperazine
C1CN(CCN1)CC2=CC(=CC=C2)F
194.253
194.25
97%
1.106 g/mL
1.529
Colorless to Orange
55513-19-4
MFCD02242870
ITHBJSRWFNLKIH-UHFFFAOYSA-N
1-[(3-fluorophenyl)methyl]piperazine
903813
Liquid

Chemical Identifiers

55513-19-4
194.253
ITHBJSRWFNLKIH-UHFFFAOYSA-N
903813
C1CN(CCN1)CC2=CC(=CC=C2)F
C11H15FN2
MFCD02242870
1-3-fluoro-benzyl-piperazine
1-[(3-fluorophenyl)methyl]piperazine
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents