Please login to your online account to display your discounted pricing

1,3-Dithiane, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC169511000

 View more versions of this product

Catalog No. AC169511000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 120.23
Physical Form Crystals, Needles or Powder
Quantity 100g
Color White to pale beige
Melting Point 49° to 54°C
Density 90g/mL
Fieser 02,182; 03,135; 04,216; 05,287; 06,248; 07,142; 14,157; 16,161
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Molecular Formula C4H8S2
Assay Percent Range 97%
CAS 505-23-7
Chemical Name or Material 1, 3-Dithiane
MDL Number MFCD00006654
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1, 3-Dithiane
CAS 505-23-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H8S2
Formula Weight 120.23
MDL Number MFCD00006654