Please login to your online account to display your discounted pricing

1,3-Dipropyl-8-phenylxanthine, Tocris

A1 selective antagonist

$162.94 - $684.37

Chemical Identifiers

Chemical Name or Material 1, 3-Dipropyl-8-phenylxanthine
Assay Percent Range >98%
Molecular Formula C17H20N4O2
Formula Weight 312.37
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

048650

 
r & d systems
048650
50mg Each for $684.37

048610

 
r & d systems
048610
10mg Each for $162.94
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 312.37
Chemical Name or Material 1, 3-Dipropyl-8-phenylxanthine
Assay Percent Range >98%
Molecular Formula C17H20N4O2
CAS 85872-53-3
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature