Please login to your online account to display your discounted pricing

1,3-Dipropyl-8-phenylxanthine, Tocris


A1 selective antagonist

Quantity 50mg

Manufacturer: r & d systems  048650

 View more versions of this product

Catalog No. 048650

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 312.37
Chemical Name or Material 1, 3-Dipropyl-8-phenylxanthine
Assay Percent Range >98%
Molecular Formula C17H20N4O2
CAS 85872-53-3
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material 1, 3-Dipropyl-8-phenylxanthine
Assay Percent Range >98%
Molecular Formula C17H20N4O2
Formula Weight 312.37
Accessories

Store at Room Temperature