missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dipropyl-8-phenylxanthine

Selective A1 adenosine antagonist

$242.11 - $874.38

Chemical Identifiers

CAS 85872-53-3
Molecular Formula C17H20N4O2
Molecular Weight (g/mol) 312.373
MDL Number MFCD00209811
InChI Key CLIGSMOZKDCDRZ-UHFFFAOYSA-N
Synonym 1,3-dipropyl-8-phenylxanthine, 3,7-dihydro-1,3-dipropyl-8-phenyl-1h-purine-2,6,dione, 8-phenyl-1,3-dipropyl-9h-purine-2,6-dione, 8-phenyl-1,3-dipropyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6,dione, 3,7-dihydro-1,3-dipropyl-8-phenyl, 1h-purine-2,6-dione, 3,7-dihydro-8-phenyl-1,3-dipropyl, tocris-0486, 8-phenyl-1,3-dipropyl-3,9-dihydro-1h-purine-2,6-dione, 8-phenyl-1,3-dipropyl-3,9-dihydro-purine-2,6-dione
PubChem CID 128784
IUPAC Name 8-phenyl-1,3-dipropyl-7H-purine-2,6-dione
SMILES CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63554MA View Documents Alfa Aesar™
J63554MA
10mg Each for $242.11
 
AAJ63554MF View Documents Alfa Aesar™
J63554MF
50mg Each for $874.38
 
Specifications

Chemical Identifiers

85872-53-3
312.373
CLIGSMOZKDCDRZ-UHFFFAOYSA-N
128784
CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C3=CC=CC=C3
C17H20N4O2
MFCD00209811
1,3-dipropyl-8-phenylxanthine, 3,7-dihydro-1,3-dipropyl-8-phenyl-1h-purine-2,6,dione, 8-phenyl-1,3-dipropyl-9h-purine-2,6-dione, 8-phenyl-1,3-dipropyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6,dione, 3,7-dihydro-1,3-dipropyl-8-phenyl, 1h-purine-2,6-dione, 3,7-dihydro-8-phenyl-1,3-dipropyl, tocris-0486, 8-phenyl-1,3-dipropyl-3,9-dihydro-1h-purine-2,6-dione, 8-phenyl-1,3-dipropyl-3,9-dihydro-purine-2,6-dione
8-phenyl-1,3-dipropyl-7H-purine-2,6-dione

Specifications

White
85872-53-3
MFCD00209811
Soluble in ethanol (5 mM), or DMSO (10 mM) with warming
CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C3=CC=CC=C3
312.373
312.37
1,3-Dipropyl-8-phenylxanthine
10mg
C17H20N4O2
1,3-dipropyl-8-phenylxanthine, 3,7-dihydro-1,3-dipropyl-8-phenyl-1h-purine-2,6,dione, 8-phenyl-1,3-dipropyl-9h-purine-2,6-dione, 8-phenyl-1,3-dipropyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6,dione, 3,7-dihydro-1,3-dipropyl-8-phenyl, 1h-purine-2,6-dione, 3,7-dihydro-8-phenyl-1,3-dipropyl, tocris-0486, 8-phenyl-1,3-dipropyl-3,9-dihydro-1h-purine-2,6-dione, 8-phenyl-1,3-dipropyl-3,9-dihydro-purine-2,6-dione
CLIGSMOZKDCDRZ-UHFFFAOYSA-N
8-phenyl-1,3-dipropyl-7H-purine-2,6-dione
128784
Powder
Safety and Handling

Safety and Handling

RTECSNumber : UO8430880