Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Diphenylpropane, 98%

$104.53 - $381.74

Chemical Identifiers

Chemical Name or Material 1,3-Diphenylpropane
CAS 1081-75-0
Molecular Formula C15H16
Linear Formula C6H5(CH2)3C6H5
Formula Weight 196.29
MDL Number MFCD00043574
Beilstein 2044541
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0884904

  SDS
alfa aesar™
L0884904
2g Each for $104.53

AAL0884909

  SDS
alfa aesar™
L0884909
10g Each for $381.74
Description & Specifications

Specifications

Formula Weight 196.29
Chemical Name or Material 1,3-Diphenylpropane
Assay 98%
Molecular Formula C15H16
CAS 1081-75-0
Linear Formula C6H5(CH2)3C6H5
MDL Number MFCD00043574
Beilstein 2044541
Boiling Point 158° to 159°C (10mmHg)
Quantity 2g

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.