Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Diphenylacetone p-toluenesulfonylhydrazone, 99%

$65.85 - $209.45

Chemical Identifiers

Chemical Name or Material 1,3-Diphenylacetone p-toluenesulfonylhydrazone
CAS 19816-88-7
Molecular Formula C22H22N2O2S
Formula Weight 378.49
MDL Number MFCD00009645
Beilstein 2225293
Synonym Dibenzyl ketone tosyl hydrazone; 1,3-Diphenyl-2-propanone p-toluenesulfonylhydrazone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0362509

  SDS
alfa aesar™
L0362509
10g Each for $65.85

AAL0362518

  SDS
alfa aesar™
L0362518
50g Each for $209.45
Description & Specifications

Specifications

Chemical Name or Material 1,3-Diphenylacetone p-toluenesulfonylhydrazone
CAS 19816-88-7
Assay 99%
Molecular Formula C22H22N2O2S
Formula Weight 378.49
MDL Number MFCD00009645
Beilstein 2225293
Synonym Dibenzyl ketone tosyl hydrazone; 1,3-Diphenyl-2-propanone p-toluenesulfonylhydrazone
Melting Point ≈185°C
Quantity 10g