missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyl-2-thiourea 98.0+%, TCI America™

Supplier:  TCI America T0197500G

 View more versions of this product

Catalog No. T0197500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,3-Diphenyl-2-thiourea
151°C
C13H12N2S
500 g
thiocarbanilide, diphenylthiourea, 1,3-diphenyl-2-thiourea, n,n'-diphenylthiourea, sulfocarbanilide, sym-diphenylthiourea, thiourea, n,n'-diphenyl, stabilisator c, rhenocure ca, vulkacit ca
S=C(NC1=CC=CC=C1)NC1=CC=CC=C1
228.31
228.31
Crystalline Powder
102-08-9
White
MFCD00004921
2811
FCSHMCFRCYZTRQ-UHFFFAOYSA-N
1,3-diphenylthiourea
700999
≥98.0% (N)

Chemical Identifiers

102-08-9
228.31
FCSHMCFRCYZTRQ-UHFFFAOYSA-N
700999
S=C(NC1=CC=CC=C1)NC1=CC=CC=C1
C13H12N2S
MFCD00004921
thiocarbanilide, diphenylthiourea, 1,3-diphenyl-2-thiourea, n,n'-diphenylthiourea, sulfocarbanilide, sym-diphenylthiourea, thiourea, n,n'-diphenyl, stabilisator c, rhenocure ca, vulkacit ca
1,3-diphenylthiourea
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2205

RTECSNumber : FE1225000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions