missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Diphenyl-2-propyn-1-ol, tech. 90%

Manufacturer:  Alfa Aesar™ H5337406

 View more versions of this product

Catalog No. AAH5337406



Description

Description

Applications
1,3-Diphenyl-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

1,3-Diphenyl-2-propyn-1-ol
5g
C15H12O
UN3082
DZZWMODRWHHWFR-UHFFFAOYSA-N
1,3-diphenylprop-2-yn-1-ol
296659
90%
>110°C (230°F)
1817-49-8
MFCD06654198
1,3-diphenyl-2-propyn-1-ol, 1,3-diphenyl-prop-2-yn-1-ol, 1-phenyl-3-phenyl-propyn-3-ol, 1,3-diphenylpropargyl alcohol, 1,3-diphenyl-2-propyn-1-ol, 1,3-diphenyl-2-propyn-1-ol, benzenemethanol, a-2-phenylethynyl, alpha-phenyl ethynyl-benzenemethanol
C1=CC=C(C=C1)C#CC(C2=CC=CC=C2)O
208.26
208.26
Technical

Chemical Identifiers

1817-49-8
208.26
DZZWMODRWHHWFR-UHFFFAOYSA-N
296659
C1=CC=C(C=C1)C#CC(C2=CC=CC=C2)O
C15H12O
MFCD06654198
1,3-diphenyl-2-propyn-1-ol, 1,3-diphenyl-prop-2-yn-1-ol, 1-phenyl-3-phenyl-propyn-3-ol, 1,3-diphenylpropargyl alcohol, 1,3-diphenyl-2-propyn-1-ol, 1,3-diphenyl-2-propyn-1-ol, benzenemethanol, a-2-phenylethynyl, alpha-phenyl ethynyl-benzenemethanol
1,3-diphenylprop-2-yn-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

SDS Documents