Please login to your online account to display your discounted pricing

1,3-Diphenoxybenzene, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC408840050

 View more versions of this product

Catalog No. AC408840050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (GC)
Formula Weight 262.3
Physical Form Crystals or Crystalline Powder
Quantity 5g
Color White
Melting Point 58° to 61°C
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C18H14O2
Chemical Name or Material 1, 3-Diphenoxybenzene
CAS 3379-38-2
Assay Percent Range 99%

Specifications

Chemical Name or Material 1, 3-Diphenoxybenzene
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C18H14O2
Formula Weight 262.3