Please login to your online account to display your discounted pricing

1,3-Diphenoxybenzene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Diphenoxybenzene
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C18H14O2
Formula Weight 262.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC408840050

  SDS
thermo fisher scientific
AC408840050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC408840250

  SDS
thermo fisher scientific
408840250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC408841000

  SDS
thermo fisher scientific
AC408841000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5% (GC)
Formula Weight 262.3
Physical Form Crystals or Crystalline Powder
Quantity 5g
Color White
Melting Point 58° to 61°C
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C18H14O2
Chemical Name or Material 1, 3-Diphenoxybenzene
CAS 3379-38-2
Assay Percent Range 99%