Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Diphenoxybenzene, 99%

$69.46 - $232.04

Chemical Identifiers

Chemical Name or Material 1,3-Diphenoxybenzene
CAS 3379-38-2
Molecular Formula C18H14O2
Formula Weight 262.31
MDL Number MFCD00003036
Beilstein 2054179
Synonym Resorcinol diphenyl ether
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1664706

 
alfa aesar™
A1664706
Each for $69.46

AAA1664714

 
alfa aesar™
A1664714
Each for $232.04
Description & Specifications

Specifications

Formula Weight 262.31
Assay 99%
TSCA TSCA
Quantity 5g
Beilstein 2054179
Synonym Resorcinol diphenyl ether
Molecular Formula C18H14O2
Melting Point 59° to 61°C
MDL Number MFCD00003036
CAS 3379-38-2
Chemical Name or Material 1,3-Diphenoxybenzene