Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Diphenoxy-2-propanol, 98+%

$38.12 - $136.50

Chemical Identifiers

Chemical Name or Material 1,3-Diphenoxy-2-propanol
CAS 622-04-8
Molecular Formula C15H16O3
Formula Weight 244.29
MDL Number MFCD00091115
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2050303

  SDS
alfa aesar™
B2050303
1g Each for $38.12

AAB2050306

  SDS
alfa aesar™
B2050306
5g Each for $136.50
Description & Specifications

Specifications

Formula Weight 244.29
RTECS Number UB4100000
Melting Point 80° to 84°C
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Chemical Name or Material 1,3-Diphenoxy-2-propanol
Health Hazard 1 Danger
Quantity 1g
CAS 622-04-8
Molecular Formula C15H16O3
MDL Number MFCD00091115
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.