Learn More
1,3-Dioxolane, 99.5%, stab., Thermo Scientific Chemicals
As a solvent, in Mannich reaction, as an electrolyte in batteries
$61.96 - $460.19
Chemical Identifiers
CAS | 646-06-0 |
---|---|
Molecular Formula | C3H6O2 |
Molecular Weight (g/mol) | 74.079 |
MDL Number | MFCD00003207 |
InChI Key | WNXJIVFYUVYPPR-UHFFFAOYSA-N |
Synonym | dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech |
PubChem CID | 12586 |
ChEBI | CHEBI:87597 |
IUPAC Name | 1,3-dioxolane |
SMILES | C1COCO1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA42897AK
|
Thermo Scientific Chemicals
042897AK |
250 mL |
Each for $61.96
|
|
|||||
AA42897K2
|
Thermo Scientific Chemicals
042897K2 |
1 L |
Each for $155.47
|
|
|||||
AA42897M1
|
Thermo Scientific Chemicals
042897M1 |
4 x 1 L |
Each for $460.19
|
|
|||||
Description
As a solvent, in Mannich reaction, as an electrolyte in batteries.1,3-Dioxolane is used as a solvent for paints, cellulose esters and in organic synthesis. It is used as an extragent for waxes, fats and oils, stabilizer for halogenated hydrocarbons and co monomer in polyacetals preparation. It is used as finishing agent and swelling agent in textiles. It acts as an intermediate for the preparation of acyclovir and vandetanib. It is an essential ingredient in industrial polymers. In polymerization reactions, it acts as a chain length regulator and a chain transfer agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
646-06-0 | |
74.079 | |
WNXJIVFYUVYPPR-UHFFFAOYSA-N | |
12586 | |
1,3-dioxolane |
C3H6O2 | |
MFCD00003207 | |
dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech | |
CHEBI:87597 | |
C1COCO1 |
Specifications
646-06-0 | |
1.06 g/mL | |
1°C (34°F) | |
1.4 | |
250 mL | |
dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech | |
WNXJIVFYUVYPPR-UHFFFAOYSA-N | |
1,3-dioxolane | |
12586 | |
74.08 | |
Liquid |
-95°C | |
74°C to 75°C | |
C3H6O2 | |
MFCD00003207 | |
UN1166 | |
Miscible with water,ethanol,ether,n-octanol,acetone,chloroform,dichloromethane,ethyl Acetate and methanol. | |
C1COCO1 | |
74.079 | |
CHEBI:87597 | |
99.5% | |
1,3-Dioxolane, Stabilized |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
H225-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIOXOLANE
EINECSNumber : 211-463-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only