Please login to your online account to display your discounted pricing

(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide, 98%, ACROS Organics™

$32.04 - $32.04

Chemical Identifiers

Chemical Name or Material (1, 3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide
CAS 52509-14-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C22H22BrO2P
Formula Weight 429.28
MDL Number MFCD00011966
Synonym Triphenyl-1, 3-dioxolan-2-ylmethylphosphoniumbromide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC208030010

  SDS
thermo fisher scientific
208030010
Glass bottle 1g Each for $32.04
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 429.28
Physical Form Powder or Crystals
Quantity 1g
Color White to light brown
Melting Point 185° to 193°C
CAS Max % 100.0
Synonym Triphenyl-1, 3-dioxolan-2-ylmethylphosphoniumbromide
Assay Percent Range 98%
Molecular Formula C22H22BrO2P
MDL Number MFCD00011966
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material (1, 3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide
CAS 52509-14-5
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.